accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NN

CAS RN 1620-76-4

IUPAC Name: 4-methylpyridine-2-carbonitrile
Synonyms: 4-methylpyridine-2-carbonitrile
Molecular Weight (g/mol): 118.14
Molecular Formula: C7H6N2
InChi Key: LQAWSWUFSHYCHP-UHFFFAOYSA-N
SMILES: CC1=CC(=NC=C1)C#N
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results