CAS RN 162148-48-3
IUPAC Name:
tert-butyl 2-{7-[2-(tert-butoxy)-2-oxoethyl]-1,4,7,10-tetraazacyclododecan-1-yl}acetate
Synonyms:
tert-butyl 2-{7-[2-(tert-butoxy)-2-oxoethyl]-1,4,7,10-tetraazacyclododecan-1-yl}acetate
Molecular Weight (g/mol):
400.56
Molecular Formula:
C20H40N4O4
InChi Key:
OERWIIJOLSXGAQ-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)CN1CCNCCN(CC(=O)OC(C)(C)C)CCNCC1