accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1622-32-8

IUPAC Name: 2-chloroethane-1-sulfonyl chloride
Synonyms: 2-chloroethanesulfonyl chloride
Molecular Weight (g/mol): 163.01
Molecular Formula: C2H4Cl2O2S
InChi Key: VHCSBTPOPKFYIU-UHFFFAOYSA-N
SMILES: ClCCS(Cl)(=O)=O
Boiling Point 
  • (4)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)