accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHOFFFFFFOH

CAS RN 16261-80-6

IUPAC Name: 4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)benzoic acid
Synonyms: 4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)benzoic acid
Molecular Weight (g/mol): 288.15
Molecular Formula: C10H6F6O3
InChi Key: DLJNNINHDYILFL-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)C(O)(C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results