accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 16263-52-8

IUPAC Name: 3-chloro-1,2-benzoxazole
Synonyms: 3-chloro-1,2-benzoxazole
Molecular Weight (g/mol): 153.57
Molecular Formula: C7H4ClNO
InChi Key: INWUFXPCLZRSBH-UHFFFAOYSA-N
SMILES: ClC1=NOC2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results