accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOOOHOOOOOOHOHOOHOOON(S)(R)(R)(R)(S)H3C(S)(R)(R)H3C(R)H3CCH3(S)H3C(R)CH3H3C(R)CH3(R)(S)CH3CH3CH3HHH(?)(?)(?)(?)

CAS RN 162635-04-3

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: —
InChi Key: —
SMILES: —
Product Type 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results