accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 16265-04-6

IUPAC Name: 2-chloro-1H-imidazole
Synonyms: 2-chloroimidazole
Molecular Weight (g/mol): 102.52
Molecular Formula: C3H3ClN2
InChi Key: OCVXSFKKWXMYPF-UHFFFAOYSA-N
SMILES: ClC1=NC=CN1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results