accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

N

CAS RN 16269-13-9

IUPAC Name: 2-(adamantan-1-yl)acetonitrile
Synonyms: 2-(adamantan-1-yl)acetonitrile
Molecular Weight (g/mol): 175.28
Molecular Formula: C12H17N
InChi Key: DXQVPXCZIRQITG-UHFFFAOYSA-N
SMILES: N#CCC12CC3CC(CC(C3)C1)C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results