accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SSN

CAS RN 16278-99-2

IUPAC Name: [2,2'-bithiophene]-5-carbonitrile
Synonyms: [2,2'-bithiophene]-5-carbonitrile
Molecular Weight (g/mol): 191.27
Molecular Formula: C9H5NS2
InChi Key: ANARYBGZNUMARH-UHFFFAOYSA-N
SMILES: N#CC1=CC=C(S1)C1=CC=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results