accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 162856-35-1

IUPAC Name: 1-(tert-butyldimethylsilyl)-4-oxoazetidine-2-carboxylic acid
Synonyms: 1-(tert-butyldimethylsilyl)-4-oxoazetidine-2-carboxylic acid
Molecular Weight (g/mol): 229.35
Molecular Formula: C10H19NO3Si
InChi Key: LIEWITJXZYCDLE-UHFFFAOYNA-N
SMILES: CC(C)(C)[Si](C)(C)N1C(CC1=O)C(O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results