accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 163008-86-4

CH3SNNOH

CAS RN 163008-86-4

IUPAC Name: (4-methyl-1,2,3-thiadiazol-5-yl)methanol
Synonyms: (4-methyl-1,2,3-thiadiazol-5-yl)methanol
Molecular Weight (g/mol): 130.17
Molecular Formula: C4H6N2OS
InChi Key: MQLRVKHKJCRSBO-UHFFFAOYSA-N
SMILES: CC1=C(CO)SN=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results