accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 16310-13-7

IUPAC Name: (2S)-1,3-thiazolidin-3-ium-2-carboxylate
Synonyms: (2S)-1,3-thiazolidin-3-ium-2-carboxylate
Molecular Weight (g/mol): 133.17
Molecular Formula: C4H7NO2S
InChi Key: ULSZVNJBVJWEJE-VKHMYHEASA-N
SMILES: [O-]C(=O)[C@H]1[NH2+]CCS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results