accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 163222-33-1

HO(S)(R)(S)NOFOHF

CAS RN 163222-33-1

IUPAC Name: (3R,4S)-1-(4-fluorophenyl)-3-[(3S)-3-(4-fluorophenyl)-3-hydroxypropyl]-4-(4-hydroxyphenyl)azetidin-2-one
Synonyms: ezetimibe zetia ezetrol ezetimib ezedoc ezetimibe (product) ezetimibe preparation ezetimibe (substance) msd brand of ezetimibe merck brand of ezetimibe
Molecular Weight (g/mol): 409.43
Molecular Formula: C24H21F2NO3
InChi Key: OLNTVTPDXPETLC-XPWALMASSA-N
SMILES: O[C@@H](CC[C@@H]1[C@H](N(C1=O)C1=CC=C(F)C=C1)C1=CC=C(O)C=C1)C1=CC=C(F)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results