accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1633-22-3

IUPAC Name: tricyclo[8.2.2.2⁴,⁷]hexadeca-1(12),4,6,10,13,15-hexaene
Synonyms: paracyclophane
Molecular Weight (g/mol): 208.3
Molecular Formula: C16H16
InChi Key: OOLUVSIJOMLOCB-UHFFFAOYSA-N
SMILES: C1CC2=CC=C(CCC3=CC=C1C=C3)C=C2
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results