accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CClO

CAS RN 16331-45-6

IUPAC Name: 4-ethylbenzoyl chloride
Synonyms: 4-ethylbenzoyl chloride
Molecular Weight (g/mol): 168.62
Molecular Formula: C9H9ClO
InChi Key: AVTLLLZVYYPGFX-UHFFFAOYSA-N
SMILES: CCC1=CC=C(C=C1)C(Cl)=O
Boiling Point 
  • (5)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (5)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)