accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHOHNNO

CAS RN 1634-82-8

IUPAC Name: 2-[2-(4-oxocyclohexa-2,5-dien-1-ylidene)hydrazin-1-yl]benzoic acid
Synonyms: 2-[2-(4-oxocyclohexa-2,5-dien-1-ylidene)hydrazin-1-yl]benzoic acid
Molecular Weight (g/mol): 242.23
Molecular Formula: C13H10N2O3
InChi Key: FBVSMDPNVYJNON-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1NN=C1C=CC(=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results