accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3OON(R)OO

CAS RN 163438-09-3

IUPAC Name: (3R)-1-[(tert-butoxy)carbonyl]piperidine-3-carboxylate
Synonyms: (3R)-1-(tert-butoxycarbonyl)piperidine-3-carboxylate
Molecular Weight (g/mol): 228.27
Molecular Formula: C11H18NO4
InChi Key: NXILIHONWRXHFA-MRVPVSSYSA-M
SMILES: CC(C)(C)OC(=O)N1CCC[C@H](C1)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)