accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3NH2NOO

CAS RN 1635-84-3

IUPAC Name: 2,4-dimethyl-6-nitroaniline
Synonyms: 2,4-dimethyl-6-nitroaniline
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: VSRYYONYIUUFFY-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(N)C(=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results