accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 16369-05-4

IUPAC Name: (2S)-1-hydroxy-3-methylbutan-2-aminium
Synonyms: valinol
Molecular Weight (g/mol): 104.17
Molecular Formula: C5H14NO
InChi Key: NWYYWIJOWOLJNR-RXMQYKEDSA-O
SMILES: CC(C)[C@H]([NH3+])CO
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results