accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1637-73-6

IUPAC Name: trisodium 1-hydroxypropane-1,2,3-tricarboxylate
Synonyms: trisodium isocitrate
Molecular Weight (g/mol): 258.07
Molecular Formula: C6H5Na3O7
InChi Key: HWMVXEKEEAIYGB-UHFFFAOYNA-K
SMILES: [Na+].[Na+].[Na+].OC(C(CC([O-])=O)C([O-])=O)C([O-])=O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results