accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 16373-93-6

IUPAC Name: 5-(6-amino-9H-purin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Synonyms: adenosine, 2'-deoxy-
Molecular Weight (g/mol): 251.25
Molecular Formula: C10H13N5O3
InChi Key: OLXZPDWKRNYJJZ-UHFFFAOYNA-N
SMILES: NC1=C2N=CN(C3CC(O)C(CO)O3)C2=NC=N1
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)