accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1638-86-4

H3COPOCH3

CAS RN 1638-86-4

IUPAC Name: diethyl phenylphosphonite
Synonyms: diethyl phenylphosphonite
Molecular Weight (g/mol): 198.2
Molecular Formula: C10H15O2P
InChi Key: RVDJLKVICMLVJQ-UHFFFAOYSA-N
SMILES: CCOP(OCC)C1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)