accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 16413-26-6

IUPAC Name: 3-isocyanatobenzonitrile
Synonyms: 3-isocyanatobenzonitrile
Molecular Weight (g/mol): 144.13
Molecular Formula: C8H4N2O
InChi Key: NZHPVPMRNASEQK-UHFFFAOYSA-N
SMILES: O=C=NC1=CC=CC(=C1)C#N
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results