accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3BHOOH

CAS RN 16419-60-6

IUPAC Name: (2-methylphenyl)boronic acid
Synonyms: 2-methylphenylboronic acid
Molecular Weight (g/mol): 135.96
Molecular Formula: C7H9BO2
InChi Key: NSJVYHOPHZMZPN-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1B(O)O
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)