accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNFFPFFFFF

CAS RN 164298-25-3

IUPAC Name: 1-[fluoro(pyrrolidin-1-yl)methylidene]-1λ⁵-pyrrolidin-1-ylium; hexafluoro-λ⁵-phosphanuide
Synonyms: 1-[fluoro(pyrrolidin-1-yl)methylidene]-1λ⁵-pyrrolidin-1-ylium hexafluorophosphate
Molecular Weight (g/mol): 316.2
Molecular Formula: C9H16F7N2P
InChi Key: MNJUGQKOHJQOCK-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.FC(N1CCCC1)=[N+]1CCCC1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results