accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1643-16-9

IUPAC Name: 2-[4-(propan-2-yl)phenoxy]acetic acid
Synonyms: P-I-propylphenoxyacetic acid
Molecular Weight (g/mol): 194.23
Molecular Formula: C11H14O3
InChi Key: FPVCSFOUVDLTDG-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(OCC(O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results