accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrH3CNCH3H3CH3C

CAS RN 1643-19-2

IUPAC Name: tetrabutylazanium bromide
Synonyms: tetrabutylammonium bromide tetra-N-butylammonium bromide TBAB
Molecular Weight (g/mol): 322.38
Molecular Formula: C16H36BrN
InChi Key: JRMUNVKIHCOMHV-UHFFFAOYSA-M
SMILES: [Br-].CCCC[N+](CCCC)(CCCC)CCCC
Molecular Weight (g/mol) 
  • (5)
  • (3)
  • (4)
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (3)
  • (5)
Physical Form 
  • (5)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (5)
  • (3)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (5)
  • (5)
  • (1)