accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNCH3H3CCH3OO

CAS RN 16436-29-6

IUPAC Name: tetrahexylazanium benzoate
Synonyms: tetrahexylazanium benzoate
Molecular Weight (g/mol): 475.8
Molecular Formula: C31H57NO2
InChi Key: PSEQWFPWQRZBOO-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=CC=C1.CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results