accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1644-68-4

IUPAC Name: 2,3-bis(trifluoromethyl)pyridine
Synonyms: 2,3-bis(trifluoromethyl)pyridine
Molecular Weight (g/mol): 215.1
Molecular Formula: C7H3F6N
InChi Key: RRNXYHYDSDAOFW-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(N=CC=C1)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results