accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 164461-18-1

IUPAC Name: (pyren-1-yl)boronic acid
Synonyms: pyren-1-ylboronic acid
Molecular Weight (g/mol): 246.07
Molecular Formula: C16H11BO2
InChi Key: MWEKPLLMFXIZOC-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results