accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 16452-01-0

IUPAC Name: 3-methoxy-4-methylaniline
Synonyms: 5-amino-2-methyl-anisol
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: ONADZNBSLRAJFW-UHFFFAOYSA-N
SMILES: COC1=CC(N)=CC=C1C
Molecular Weight (g/mol) 
  • (3)
  • (9)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (3)
  • (7)
  • (3)
  • (9)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (7)