accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3CH3BrH3C

CAS RN 1646-53-3

IUPAC Name: 3-bromo-1,2,4,5-tetramethylbenzene
Synonyms: 3-bromo-1,2,4,5-tetramethylbenzene
Molecular Weight (g/mol): 213.12
Molecular Formula: C10H13Br
InChi Key: WJKBPTLQJXKEHC-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C)C(Br)=C1C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results