accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 164670-44-4

IUPAC Name: (pyridin-4-yl)thiourea
Synonyms: pyridin-4-ylthiourea
Molecular Weight (g/mol): 153.2
Molecular Formula: C6H7N3S
InChi Key: HLOFIQOOOSRNFY-UHFFFAOYSA-N
SMILES: NC(=S)NC1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results