accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FNClHNNN

CAS RN 1651-29-2

IUPAC Name: 6-chloro-2-fluoro-7H-purine
Synonyms: 6-chloro-2-fluoropurine
Molecular Weight (g/mol): 172.55
Molecular Formula: C5H2ClFN4
InChi Key: UNRIYCIDCQDGQE-UHFFFAOYSA-N
SMILES: FC1=NC(Cl)=C2NC=NC2=N1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results