accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2NH2OHHO

CAS RN 16523-31-2

IUPAC Name: 4,6-diaminobenzene-1,3-diol
Synonyms: 4,6-diaminobenzene-1,3-diol
Molecular Weight (g/mol): 140.14
Molecular Formula: C6H8N2O2
InChi Key: DPYROBMRMXHROQ-UHFFFAOYSA-N
SMILES: NC1=CC(N)=C(O)C=C1O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results