CAS RN 1653-31-2
CAS RN 1653-31-2
IUPAC Name:
tridecan-2-ol
Synonyms:
2-tridecanol
Molecular Weight (g/mol):
200.37
Molecular Formula:
C13H28O
InChi Key:
HKOLRKVMHVYNGG-UHFFFAOYNA-N
SMILES:
CCCCCCCCCCCC(C)O
1
–
1
of
1
results
1
2-Tridecanol, 97%
CAS: 1653-31-2 Molecular Formula: C13H28O Molecular Weight (g/mol): 200.37 MDL Number: MFCD00037601 InChI Key: HKOLRKVMHVYNGG-UHFFFAOYNA-N Synonym: 2-tridecanol,2-tridecyl alcohol,2-hydroxytridecane,methylundecylcarbinol,2-tridecanol, s,alcohols, c11-15-secondary,linear secondary c11-c15 alcohol,acmc-20m48e,acmc-1c00w PubChem CID: 15449 IUPAC Name: tridecan-2-ol SMILES: CCCCCCCCCCCC(C)O