accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaNaOSOOSOOO

CAS RN 1655-43-2

IUPAC Name: disodium naphthalene-1,6-disulfonate
Synonyms: disodium naphthalene-1,6-disulfonate
Molecular Weight (g/mol): 332.25
Molecular Formula: C10H6Na2O6S2
InChi Key: FXJFYEOXUWERCL-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2C(C=CC=C2S([O-])(=O)=O)=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results