accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1655-45-4

IUPAC Name: disodium naphthalene-2,6-disulfonate
Synonyms: disodium 2,6-naphthalenedisulfonate
Molecular Weight (g/mol): 332.25
Molecular Formula: C10H6Na2O6S2
InChi Key: WZZLWPIYWZEJOX-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC2=CC=C(C=C2C=C1)S([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results