accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 165534-43-0

CH3OPOOCH3ONNNO

CAS RN 165534-43-0

IUPAC Name: diethyl 4-oxo-3,4-dihydro-1,2,3-benzotriazin-3-yl phosphate
Synonyms: depbt
Molecular Weight (g/mol): 299.22
Molecular Formula: C11H14N3O5P
InChi Key: AJDPNPAGZMZOMN-UHFFFAOYSA-N
SMILES: CCOP(=O)(OCC)ON1N=NC2=CC=CC=C2C1=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)