CAS RN 16555-77-4
IUPAC Name:
(2R)-2-hydroxy-2-(phenylformamido)acetate
Synonyms:
(2R)-2-hydroxy-2-(phenylformamido)acetate
Molecular Weight (g/mol):
194.17
Molecular Formula:
C9H8NO4
InChi Key:
GCWCVCCEIQXUQU-MRVPVSSYSA-M
SMILES:
O[C@@H](NC(=O)C1=CC=CC=C1)C([O-])=O