accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 16576-63-9

IUPAC Name: niobium(5+) pentabenzenolate
Synonyms: niobium(5+) pentaphenolate
Molecular Weight (g/mol): 558.43
Molecular Formula: C30H25NbO5
InChi Key: ZCNDFHXTXLMYDM-UHFFFAOYSA-I
SMILES: [Nb+5].[O-]C1=CC=CC=C1.[O-]C1=CC=CC=C1.[O-]C1=CC=CC=C1.[O-]C1=CC=CC=C1.[O-]C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results