accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 165800-06-6

IUPAC Name: [1-hydroxy-2-(1H-imidazol-1-yl)-1-phosphonoethyl]phosphonic acid hydrate
Synonyms: hydrate zoledronate
Molecular Weight (g/mol): 290.11
Molecular Formula: C5H12N2O8P2
InChi Key: FUXFIVRTGHOMSO-UHFFFAOYSA-N
SMILES: O.OC(CN1C=CN=C1)(P(O)(O)=O)P(O)(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results