accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 166108-71-0

IUPAC Name: 2-{2-[2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)ethoxy]ethoxy}acetic acid
Synonyms: [2-(2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}ethoxy)ethoxy]acetic acid
Molecular Weight (g/mol): 385.42
Molecular Formula: C21H23NO6
InChi Key: XQPYRJIMPDBGRW-UHFFFAOYSA-N
SMILES: OC(=O)COCCOCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (11)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (8)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (2)
  • (3)
  • (8)
  • (11)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (8)
  • (2)