accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFBrOH

CAS RN 166281-37-4

IUPAC Name: 2-bromo-4,5-difluorophenol
Synonyms: 2-bromo-4,5-difluorophenol
Molecular Weight (g/mol): 208.99
Molecular Formula: C6H3BrF2O
InChi Key: FCYZOOHWUOEAOX-UHFFFAOYSA-N
SMILES: OC1=CC(F)=C(F)C=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results