accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOBOHOHO

CAS RN 166316-48-9

IUPAC Name: 3-[4-(dihydroxyboranyl)phenyl]propanoic acid
Synonyms: 3-[4-(dihydroxyboranyl)phenyl]propanoic acid
Molecular Weight (g/mol): 193.99
Molecular Formula: C9H11BO4
InChi Key: VPSARXNVXCRDIV-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(CCC(O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results