accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BFFFK

CAS RN 166328-07-0

IUPAC Name: potassium trifluoro(naphthalen-1-yl)boranuide
Synonyms: potassium trifluoro(naphthalen-1-yl)boranuide
Molecular Weight (g/mol): 234.07
Molecular Formula: C10H7BF3K
InChi Key: SBEPDIKXYGSGJL-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C1=C2C=CC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results