accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HNSOOBr

CAS RN 166338-06-3

IUPAC Name: 3-bromo-N-phenylbenzene-1-sulfonamide
Synonyms: 3-bromo-N-phenylbenzenesulfonamide
Molecular Weight (g/mol): 312.18
Molecular Formula: C12H10BrNO2S
InChi Key: JPSZUSKQOMHYRI-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=C1)S(=O)(=O)NC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results