CAS RN 16634-88-1
IUPAC Name:
5-bromo-2-hydroxy-3-nitrobenzaldehyde
Synonyms:
5-bromo-2-hydroxy-3-nitrobenzaldehyde
Molecular Weight (g/mol):
246.02
Molecular Formula:
C7H4BrNO4
InChi Key:
YEYPSUQQZNDKDE-UHFFFAOYSA-N
SMILES:
OC1=C(C=C(Br)C=C1C=O)[N+]([O-])=O
1
–
2
of
2
results
1
5-Bromo-3-nitrosalicylaldehyde, 97%
CAS: 16634-88-1 Molecular Formula: C7H4BrNO4 Molecular Weight (g/mol): 246.016 MDL Number: MFCD00130110 InChI Key: YEYPSUQQZNDKDE-UHFFFAOYSA-N Synonym: 5-bromo-2-hydroxy-3-nitrobenzaldehyde,5-bromo-3-nitrosalicylaldehyde,benzaldehyde, 5-bromo-2-hydroxy-3-nitro,3-nitro-5-bromosalicylaldehyde,benzaldehyde,5-bromo-2-hydroxy-3-nitro,salicylaldehyde, 5-bromo-3-nitro,acmc-209du3,5-bromo-3-nitrosalicyladehyde,2-hydroxy-3-nitro-5-bromobenzaldehyde PubChem CID: 618699 SMILES: C1=C(C=C(C(=C1[N+](=O)[O-])O)C=O)Br