CAS RN 1664-40-0
IUPAC Name:
N1-phenylethane-1,2-diamine
Synonyms:
N1-phenylethane-1,2-diamine
Molecular Weight (g/mol):
136.2
Molecular Formula:
C8H12N2
InChi Key:
OCIDXARMXNJACB-UHFFFAOYSA-N
SMILES:
NCCNC1=CC=CC=C1
1
–
1
of
1
results
1
N-Phenylethylenediamine, 99%
CAS: 1664-40-0 Molecular Formula: C8H12N2 Molecular Weight (g/mol): 136.2 MDL Number: MFCD00008162 InChI Key: OCIDXARMXNJACB-UHFFFAOYSA-N Synonym: n-phenylethylenediamine,n1-phenylethane-1,2-diamine,n-2-aminoethyl aniline,1,2-ethanediamine, n-phenyl,ethylenediamine, n-phenyl,n-phenyl-1,2-ethanediamine,benzenamine, n-2-aminoethyl,n-phenylethane-1,2-diamine,1,2-ethanediamine, n1-phenyl,2-aminoethyl phenylamine PubChem CID: 74270 IUPAC Name: N'-phenylethane-1,2-diamine SMILES: C1=CC=C(C=C1)NCCN