accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 16652-64-5

(S)ONH2OHO

CAS RN 16652-64-5

IUPAC Name: 2-amino-3-[4-(benzyloxy)phenyl]propanoic acid
Synonyms: 2-amino-3-[4-(benzyloxy)phenyl]propanoic acid
Molecular Weight (g/mol): 271.32
Molecular Formula: C16H17NO3
InChi Key: KAFHLONDOVSENM-UHFFFAOYNA-N
SMILES: NC(CC1=CC=C(OCC2=CC=CC=C2)C=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)